Få mere at vide
R&D Systems™ Human Kallikrein 5 DuoSet ELISA
Beskrivelse
Indeholder
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human Kallikrein 5 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Kallikrein 5.
Tekniske data
Tekniske data
| Produkttype | ELISA Kit |
| Arter | Human |
| Bindende mål | Kallikrein-5 |
| Format | 96-brønds plader |
| Til brug med (applikation) | ELISA |
| Gen-id (Entrez) | 25818 |
| Prøvetype | Cell Culture Supernatant, Serum, Plasma |
| Indhold og opbevaring | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Research Use Only.