Få mere at vide
R&D Systems™ Human Kallikrein 5 DuoSet ELISA
Artikelnummer. 30001792
Skift visning
Format:
96-brønds plader
Artikelnummer. Format
30001792 96-brønds plader
1 options
Artikelnummer. 30001792 Leverandør R&D Systems™ Leverandørnr. DY845905
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
For the determination of natural and recombinant Kallikrein 5 in human. R&D Systems Human Kallikrein 5 DuoSet ELISA Kit is a solid phase sandwich ELISA suitable for the analysis of cell culture supernate, serum, and plasma samples.
The Human Kallikrein 5 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Kallikrein 5.
TRUSTED_SUSTAINABILITY
Tekniske data
Produkttype ELISA Kit
Arter Human
Bindende mål Kallikrein-5
Format 96-brønds plader
Til brug med (applikation) ELISA
Gen-id (Entrez) 25818
Prøvetype Cell Culture Supernatant, Serum, Plasma
Indhold og opbevaring Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Research Use Only.