Learn More
Abnova™ Human JARID2 Partial ORF (NP_004964, 1130 a.a. - 1229 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003720-Q01.25ug
Additional Details : Weight : 0.00010kg
Beschreibung
This gene is an ortholog of the mouse jumonji gene, which encodes a nuclear protein essential for mouse embryogenesis, including neural tube formation. Overexpression of mouse jumonji negatively regulates cell proliferation. The jumonji proteins contain a DNA-binding domain, called an AT-rich interaction domain (ARID), and share regions of similarity with human retinoblastoma-binding protein-2 and the human SMCX protein. [provided by RefSeq]
Sequence: LQLETSERRCQICQHLCYLSMVVQENENVVFCLECALRHVEKQKSCRGLKLMYRYDEEQIISLVNQICGKVSGKNGSIENCLSKPTPKRGPRKRATVDVPSpezifikation
NP_004964 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LQLETSERRCQICQHLCYLSMVVQENENVVFCLECALRHVEKQKSCRGLKLMYRYDEEQIISLVNQICGKVSGKNGSIENCLSKPTPKRGPRKRATVDVP | |
RUO | |
JARID2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3720 | |
JARID2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
JMJ | |
JARID2 | |
Recombinant | |
wheat germ expression system |