missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ITPKB Partial ORF (NP_002212, 545 a.a. - 643 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003707-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137041

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this protein regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of this encoded protein is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. [provided by RefSeq]

Sequence: PELLPQDQDKPFLRKACSPSNIPAVIITDMGTQEDGALEETQGSPRGNLPLRKLSSSSASSTGFSSSYEDSEEDISSDPERTLDPNSAFLHTLDQQKPR
Specifications

Specifications

NP_002212
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PELLPQDQDKPFLRKACSPSNIPAVIITDMGTQEDGALEETQGSPRGNLPLRKLSSSSASSTGFSSSYEDSEEDISSDPERTLDPNSAFLHTLDQQKPR
RUO
ITPKB
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3707
ITPKB (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IP3K/IP3K-B/IP3KB/PIG37
ITPKB
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers