Få mere at vide
Invitrogen™ Human IRGM Synthetic Peptide
Mængde:
50 μg
Pakningsstørrelse:
50µg
Beskrivelse
IRGM Synthetic Peptide, PEP-0628, from Invitrogen.
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. Two of the strongest hits implicate genes IRGM and ATG16L1, which encode proteins thought to be critical to the autophagy pathway and being significantly associated with Crohn's disease. In mouse, IRGM belongs to a family of -gamma-interferon-induced GTP-binding proteins of approximately 48 kDa. Murine IRGM induces autophagy and generates large autolysosomal organelles as a mechanism for the elimination of intracellular Mycobacterium tuberculosis. Human IRGM is also involved in autophagy and plays a role in the control of intracellular pathogens and in the reduction of intracellular bacillary load.
Tekniske data
Tekniske data
| Gen-id (Entrez) | 345611 |
| Arter | Human |
| Oprensningsmetode | purified |
| Adgangsnummer | A1A4Y4 |
| Fælles navn | Human IRGM |
| Koncentration | 200 μg/mL |
| Indhold og opbevaring | -20°C |
| Oversigt | Synthetic Peptide |
| Format | Liquid |
| Formulering | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Vis mere |