Få mere at vide
R&D Systems™ Human Integrin beta 5 (NP_002204) VersaClone cDNA
Artikelnummer. 18776809
Skift visning
Mængde:
10 μg
Artikelnummer. Mængde
18776809 10 μg
1 options
Artikelnummer. 18776809 Leverandør R&D Systems™ Leverandørnr. RDC0939
  • ONLINE EXCLUSIVE PRICE
    5975.00 DKK
    5649.00 DKK / 10µg
    Spare 326.00 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 21-10-2026
Integrin, beta 5
The Human Integrin beta 5 (NP_002204) VersaClone cDNA clone contains the complete ORF for Integrin beta 5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Integrin beta 5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Ordreinfo
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Tekniske data
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Mængde 10 μg
For Reasearch Use Only; Not for Use in Humans