missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human IL-29 (IFN lambda-1), AOF Recombinant Protein

Artikelnummer. 17606535
Klik for at se tilgængelige muligheder
Mængde:
1 mg
100 μg
500 μg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 17606535

Brand: Invitrogen™ RP87712

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

E. coli Recombinant Protein

IFN-lambdas constitute a novel family of interferons that mediate the induction of anti-viral protection in a wide variety of cells. There are three members in this family, namely lambda1, lambda2, and lambda3, which are also known as IL-29, IL-28A, and IL-28B, respectively. Similar to type I IFNs, IFN-lambdas activate an intracellular signaling to promote gene expression. IFN-lambdas mediate their anti-viral protection through a class II cytokine receptor complex composed of two essential receptor proteins, CRF2-12/IFN-lambdaR1, which is unique to IFN-lambdas, and CFR2-4/IL-10R2, which is shared with IL-10, IL-22, and IL-26 receptors. The IFN-lambdas, IL-28 and IL-29, have recently been reported to prime dendritic cells to induce proliferation of Foxp3-bearing regulatory T cells. DCs that differentiate in response to IFN-lambdas express high levels of class I and II MHC gene products, but low levels of costimulatory molecules. Moreover, these cells can specifically induce IL-2-dependent proliferation of CD4+CD25+FOXP3+ T cells with contact-dependent suppressive activity.
  • Purity: >95% by SDS-Page
  • Endotoxin Concentration: AX41 ≤EUs/μg
  • Protein Length: 179 aa.
  • Molecular Weight: 19.9 kDa
  • Animal Origin Free: Yes
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer Q8IU54
Til brug med (applikation) Control, ELISA, Gel Shift, Western Blot
Formulering 0.1% TFA with no preservative
Gen-id (Entrez) 282618
Molekylvægt (g/mol) 19.9 kDa
Navn Human IL-29, AOF
Mængde 500 μg
Kilde E. coli
Opbevaringskrav -20°C
Regulatorisk status RUO
Gene Alias Cytokine Zcyto21; FNL1; IFN lambda1; IFNL1; IFN-lambda-1; IL29; IL-29; ILN; interferon lambda 1; Interferon lambda1; Interferon lambda-1; interferon, lambda 1; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); Interleukin-29; ZCYTO21
Gen symbol IFNL1
Specifik reaktivitet Human
Konjugeret Unconjugated
Rekombinant Recombinant
Form Lyophilized
Renhed eller kvalitetsklasse >95%
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.