Learn More
Invitrogen™ Human IL-21 Proquantum Immunoassay Kit
ProQuantum Immunoassay Kit
Brand:  Invitrogen™ A355935
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 17194699
  • 25170.00 DKK / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
Human IL-21 Proquantum Immunoassay Kit
ProQuantum Immunoassay Kit
Specifications
Specifications
Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5°C to -30°C.
5 x 96 Tests