Learn More
Invitrogen™ Human IL-21 Proquantum Immunoassay Kit
ProQuantum Immunoassay Kit
Brand: Invitrogen™ A35593
Additional Details : Weight : 0.22720kg
Description
Human IL-21 Proquantum Immunoassay Kit
ProQuantum Immunoassay Kit
Specifications
Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5°C to -30°C. | |
96 Tests |