missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human IL-16 Synthetic Peptide
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
Beskrivelse
IL-16 Synthetic Peptide, PEP-0788, from Invitrogen.
The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
Tekniske data
Tekniske data
| Gen-id (Entrez) | 3603 |
| Arter | Human |
| Oprensningsmetode | purified |
| Adgangsnummer | Q14005 |
| Fælles navn | Human IL-16 |
| Koncentration | 200 μg/mL |
| Indhold og opbevaring | -20°C |
| Oversigt | Synthetic Peptide |
| Format | Liquid |
| Formulering | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion