Learn More
Abnova™ Human IKBKAP Partial ORF (NP_003631, 1242 a.a. - 1331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008518-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a scaffold protein and a regulator for 3 different kinases involved in proinflammatory signaling. This encoded protein can bind NF-kappa-B-inducing kinase (NIK) and IKKs through separate domains and assemble them into an active kinase complex. Mutations in this gene have been associated with familial dysautonomia. [provided by RefSeq]
Sequence: VLFLFEFDEQGRELQKAFEDTLQLMERSLPEIWTLTYQQNSATPVLGPNSTANSIMASYQQQKTSVPVLDAELFIPPKINRRTQWKLSLLSpecifications
NP_003631 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VLFLFEFDEQGRELQKAFEDTLQLMERSLPEIWTLTYQQNSATPVLGPNSTANSIMASYQQQKTSVPVLDAELFIPPKINRRTQWKLSLL | |
RUO | |
IKBKAP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8518 | |
IKBKAP (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781H1425/DYS/ELP1/FD/FLJ12497/IKAP/IKI3/TOT1 | |
IKBKAP | |
Recombinant | |
wheat germ expression system |