missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Human IGFBP-rP10 (NP_112191) VersaClone cDNA

Artikelnummer. 18781009
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18781009 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18781009 Leverandør R&D Systems™ Leverandørnr. RDC2504

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Human IGFBP-rP10 (NP_112191) VersaClone cDNA clone contains the complete ORF for IGFBP-rP10/KAZALD1, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The IGFBP-rP10/KAZALD1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY

Tekniske data

Gene hIGFBP-rP10
Adgangsnummer NP_112191
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gen-id (Entrez) 81621
Indhold og opbevaring Store the unopened product at -20 to -80°C
Arter Human
Mængde 10 μg
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.