missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human IDH2 Synthetic Peptide
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
Beskrivelse
IDH2 Synthetic Peptide, PEP-1091, from Invitrogen.
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NADas the electron acceptor and the other NADP. Five isocitrate dehydrogenases have been reported: three NAD-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP-dependent isocitrate dehydrogenase found in the mitochondria. It plays a role in intermediary metabolism and energy production. This protein may tightly associate or interact with the pyruvate dehydrogenase complex.
Spécification
Spécification
| Gen-id (Entrez) | 3418 |
| Arter | Human |
| Oprensningsmetode | purified |
| Adgangsnummer | P48735 |
| Fælles navn | Human IDH2 |
| Koncentration | 200 μg/mL |
| Indhold og opbevaring | -20°C |
| Oversigt | Synthetic Peptide |
| Format | Liquid |
| Formulering | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu