missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HYAL2 Partial ORF (NP_003764, 340 a.a. - 439 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008692-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179055

  • 4040.00 DKK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein which is similar in structure to hyaluronidases. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. Varying functions have been described for this protein. It has been described as a lysosomal hyaluronidase which is active at a pH below 4 and specifically hydrolyzes high molecular weight hyaluronan. It has also been described as a GPI-anchored cell surface protein which does not display hyaluronidase activity but does serve as a receptor for the oncogenic virus Jaagsiekte sheep retrovirus. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. This gene encodes two alternatively spliced transcript variants which differ only in the 5' UTR. [provided by RefSeq]

Sequence: CQYLKDYLTRLLVPYVVNVSWATQYCSRAQCHGHGRCVRRNPSASTFLHLSTNSFRLVPGHAPGEPQLRPVGELSWADIDHLQTHFRCQCYLGWSGEQCQ
Specifications

Specifications

NP_003764
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CQYLKDYLTRLLVPYVVNVSWATQYCSRAQCHGHGRCVRRNPSASTFLHLSTNSFRLVPGHAPGEPQLRPVGELSWADIDHLQTHFRCQCYLGWSGEQCQ
RUO
HYAL2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8692
HYAL2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LUCA2/LuCa-2
HYAL2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers