Learn More
Abnova™ Human HSPA1L Partial ORF (NP_005518, 561 a.a. - 641 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003305-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a 70kDa heat shock protein. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which also encode isoforms of the 70kDa heat shock protein. [provided by RefSeq]
Sequence: KGKISESDKNKILDKCNELLSWLEVNQLAEKDEFDHKRKELEQMCNPIITKLYQGGCTGPACGTGYVPGRPATGPTIEEVDSpecifications
NP_005518 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.65kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KGKISESDKNKILDKCNELLSWLEVNQLAEKDEFDHKRKELEQMCNPIITKLYQGGCTGPACGTGYVPGRPATGPTIEEVD | |
RUO | |
HSPA1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3305 | |
HSPA1L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSP70-1L/HSP70-HOM/HSP70T/hum70t | |
HSPA1L | |
Recombinant | |
wheat germ expression system |