Learn More
Abnova™ Human HRBL Full-length ORF (ENSP00000262935, 1 a.a. - 155 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | ENSP00000262935 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3268 |
Molecular Weight (g/mol) | 43.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16189604
|
Abnova™
H00003268-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16179604
|
Abnova™
H00003268-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the HIV-1 Rev binding protein (HRB) family and encodes a protein with one Arf-GAP zinc finger domain, several phe-gly (FG) motifs, and four asn-pro-phe (NPF) motifs. This protein interacts with Eps15 homology (EH) domains and plays a role in the Rev export pathway, which mediates the nucleocytoplasmic transfer of proteins and RNAs. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. The 3' UTR of this gene contains an insulin receptor substrate 3-like pseudogene. [provided by RefSeq]
Sequence: MVMAAKKGPGPGGGVSGGKAEAEAASEVWCRRVRELGGCSQAGNRHCFECAQRGVTYVDITVGSFVCTTCSGLLRGLNPPHRVKSISMTTFTEPEVVFLQSRGNEVCRKIWLGLFDARTSLVPDSRDPQKVKEFLQEKYEKKRWPDTFPRRLCQLSpecifications
ENSP00000262935 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HRBL/RABR | |
AGFG2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3268 | |
HRBL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
AGFG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |