missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HPCAL1 Full-length ORF (NP_002140.2, 1 a.a. - 193 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_002140.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3241 |
Molecular Weight (g/mol) | 48.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16179584
|
Abnova™
H00003241-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16169584
|
Abnova™
H00003241-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of neuron-specific calcium-binding proteins family found in the retina and brain. It is highly similar to human hippocalcin protein and nearly identical to the rat and mouse hippocalcin like-1 proteins. It may be involved in the calcium-dependent regulation of rhodopsin phosphorylation and may be of relevance for neuronal signalling in the central nervous system. There are two alternatively spliced transcript variants of this gene, with multiple polyadenylation sites. [provided by RefSeq]
Sequence: MGKQNSKLRPEVLQDLRENTEFTDHELQEWYKGFLKDCPTGHLTVDEFKKIYANFFPYGDASKFAEHVFRTFDTNGDGTIDFREFIIALSVTSRGKLEQKLKWAFSMYDLDGNGYISRSEMLEIVQAIYKMVSSVMKMPEDESTPEKRTDKIFRQMDTNNDGKLSLEEFIRGAKSDPSIVRLLQCDPSSASQFSpecifications
NP_002140.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGKQNSKLRPEVLQDLRENTEFTDHELQEWYKGFLKDCPTGHLTVDEFKKIYANFFPYGDASKFAEHVFRTFDTNGDGTIDFREFIIALSVTSRGKLEQKLKWAFSMYDLDGNGYISRSEMLEIVQAIYKMVSSVMKMPEDESTPEKRTDKIFRQMDTNNDGKLSLEEFIRGAKSDPSIVRLLQCDPSSASQF | |
RUO | |
HPCAL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
3241 | |
HPCAL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BDR1/HLP2/VILIP-3 | |
HPCAL1 | |
Recombinant | |
wheat germ expression system |