missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HOXB5 Full-length ORF (NP_002138.1, 1 a.a. - 269 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003215-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136311

  • 4040.00 DKK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in lung and gut development. Increased expression of this gene is associated with a distinct biologic subset of acute myeloid leukemia (AML) and the occurrence of bronchopulmonary sequestration (BPS) and congenital cystic adenomatoid malformation (CCAM) tissue. [provided by RefSeq]

Sequence: MSSYFVNSFSGRYPNGPDYQLLNYGSGSSLSGSYRDPAAMHTGSYGYNYNGMDLSVNRSSASSSHFGAVGESSRAFPAPAQEPRFRQAASSCSLSSPESLPCTNGDSHGAKPSASSPSDQATSASSSANFTEIDEASASSEPEEAASQLSSPSLARAQPEPMATSTAAPEGQTPQIFPWMRKLHISHDMTGPDGKRARTAYTRYQTLELEKEFHFNRYLTRRRRIEIAHALCLSERQIKIWFQNRRMKWKKDNKLKSMSLATAGSAFQP
Specifications

Specifications

NP_002138.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HHO.C10/HOX2/HOX2A/HU-1/Hox2.1
HOXB5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3215
HOXB5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSYFVNSFSGRYPNGPDYQLLNYGSGSSLSGSYRDPAAMHTGSYGYNYNGMDLSVNRSSASSSHFGAVGESSRAFPAPAQEPRFRQAASSCSLSSPESLPCTNGDSHGAKPSASSPSDQATSASSSANFTEIDEASASSEPEEAASQLSSPSLARAQPEPMATSTAAPEGQTPQIFPWMRKLHISHDMTGPDGKRARTAYTRYQTLELEKEFHFNRYLTRRRRIEIAHALCLSERQIKIWFQNRRMKWKKDNKLKSMSLATAGSAFQP
RUO
HOXB5
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers