Få mere at vide
Invitrogen™ Human HOXB1 (aa 60-131) Control Fragment Recombinant Protein
Beskrivelse
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-144768 (PA5-144768, PA5-111202 (PA5-111202. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Tekniske data
Tekniske data
| Adgangsnummer | P14653 |
| Koncentration | ≥5.0 mg/mL |
| Til brug med (applikation) | Blocking Assay, Control |
| Formulering | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-id (Entrez) | 3211 |
| Navn | Human HOXB1 (aa 60-131) Control Fragment |
| Mængde | 100 μL |
| Regulatorisk status | RUO |
| Gene Alias | HCFP3; homeo box B1; homeobox B1; homeobox protein Hox-2.9; Homeobox protein Hox-2 I; homeobox protein Hox-B1; HO x 2; Hox-2.9; Hox-29; HO x 2 I; Hoxb1; Hoxb-1 |
| Fælles navn | HOXB1 |
| Vis mere |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.