Learn More
Abnova™ Human HNRPAB Partial ORF (NP_004490, 186 a.a. - 285 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003182-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are produced by RNA polymerase II and are components of the heterogeneous nuclear RNA (hnRNA) complexes. They are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene, which binds to one of the components of the multiprotein editosome complex, has two repeats of quasi-RRM (RNA recognition motif) domains that bind to RNAs. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: PMDPKLNKRRGFVFITFKEEEPVKKVLEKKFHTVSGSKCEIKVAQPKEVYQQQQYGSGGRGNRNRGNRGSGGGGGGGGQGSTNYGKSQRRGGHQNNYKPYSpecifications
NP_004490 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PMDPKLNKRRGFVFITFKEEEPVKKVLEKKFHTVSGSKCEIKVAQPKEVYQQQQYGSGGRGNRNRGNRGSGGGGGGGGQGSTNYGKSQRRGGHQNNYKPY | |
RUO | |
HNRNPAB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3182 | |
HNRPAB (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABBP1/FLJ40338/HNRPAB | |
HNRNPAB | |
Recombinant | |
wheat germ expression system |