missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HIPK2 Partial ORF (AAG41236.1, 961 a.a. - 1065 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAG41236.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 28996 |
Molecular Weight (g/mol) | 37.29kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16065455
|
Abnova™
H00028996-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075455
|
Abnova™
H00028996-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HIPK2 is a conserved serine/threonine nuclear kinase that interacts with homeodomain transcription factors.[supplied by OMIM]
Sequence: TGNPRTIIVPPLKTQASEVLVECDSLVPVNTSHHSSSYKSKSSSNVTSTSGHSSGSSSGAITYRQQRPGPHFQQQQPLNLSQAQQHITTDRTGSHRRQQAYITPTSpecifications
AAG41236.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686K02111/FLJ23711/PRO0593 | |
HIPK2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
28996 | |
HIPK2 (Human) Recombinant Protein (Q01) | |
TGNPRTIIVPPLKTQASEVLVECDSLVPVNTSHHSSSYKSKSSSNVTSTSGHSSGSSSGAITYRQQRPGPHFQQQQPLNLSQAQQHITTDRTGSHRRQQAYITPT | |
RUO | |
HIPK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |