missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HIPK1 Partial ORF (AAH28408, 330 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00204851-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132437

  • 2665.00 DKK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms. [provided by RefSeq]

Sequence: CTPLMVATLHPQVATITPQYAVPFTLSCAAGRPALVEQTAAVLQAWPGGTQQILLPSTWQQLPGVALHNSVQPTAMIPEAMGSGQQLADWRNAHSHGNQYS
Specifications

Specifications

AAH28408
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CTPLMVATLHPQVATITPQYAVPFTLSCAAGRPALVEQTAAVLQAWPGGTQQILLPSTWQQLPGVALHNSVQPTAMIPEAMGSGQQLADWRNAHSHGNQYS
RUO
HIPK1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
204851
HIPK1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0630/MGC26642/MGC33446/MGC33548/Myak/Nbak2
HIPK1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers