Learn More
Abnova™ Human HBG1 Partial ORF (NP_000550, 44 a.a. - 106 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003047-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The gamma globin genes (HBG1 and HBG2) are normally expressed in the fetal liver, spleen and bone marrow. Two gamma chains together with two alpha chains constitute fetal hemoglobin (HbF) which is normally replaced by adult hemoglobin (HbA) at birth. In some beta-thalassemias and related conditions, gamma chain production continues into adulthood. The two types of gamma chains differ at residue 136 where glycine is found in the G-gamma product (HBG2) and alanine is found in the A-gamma product (HBG1). The former is predominant at birth. The order of the genes in the beta-globin cluster is: 5'-epsilon -- gamma-G -- gamma-A -- delta -- beta--3'. [provided by RefSeq]
Sequence: DSFGNLSSASAIMGNPKVKAHGKKVLTSLGDATKHLDDLKGTFAQLSELHCDKLHVDPENFKLSpecifications
NP_000550 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.67kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DSFGNLSSASAIMGNPKVKAHGKKVLTSLGDATKHLDDLKGTFAQLSELHCDKLHVDPENFKL | |
RUO | |
HBG1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3047 | |
HBG1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HBGA/HBGR/HSGGL1/PRO2979 | |
HBG1 | |
Recombinant | |
wheat germ expression system |