missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GYPB Partial ORF (NP_002091.2, 22 a.a. - 59 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002994-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179104

  • 4040.00 DKK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Glycophorins A (GYPA) and B (GYPB) are major sialoglycoproteins of the human erythrocyte membrane which bear the antigenic determinants for the MN and Ss blood groups. GYPB gene consists of 5 exons and has 97% sequence homology with GYPA from the 5' UTR to the coding sequence encoding the first 45 amino acids. In addition to the M or N and S or s antigens, that commonly occur in all populations, about 40 related variant phenotypes have been identified. These variants include all the variants of the Miltenberger complex and several isoforms of Sta; also, Dantu, Sat, He, Mg, and deletion variants Ena, S-s-U- and Mk. Most of the variants are the result of gene recombinations between GYPA and GYPB. [provided by RefSeq]

Sequence: TTEVAMHTSTSSSVTKSYISSQTNGETGQLVHRFTVPA
Specifications

Specifications

NP_002091.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
29.81kDa
12.5% SDS-PAGE Stained with Coomassie Blue
TTEVAMHTSTSSSVTKSYISSQTNGETGQLVHRFTVPA
RUO
GYPB
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2994
GYPB (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD235b/GPB/GPB.NY/GYPHe.NY/HGpMiVI/MNS/SS
GYPB
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers