Få mere at vide
R&D Systems™ Human Granzyme B DuoSet ELISA
Mængde:
1 Kit (for 5 Plates)
Beskrivelse
The Human Granzyme B DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Granzyme B.
Tekniske data
Tekniske data
| Analyseområde | 39.10 to 2500 pg/mL |
| Konjugeret | Biotin |
| Immunoassay-kitformat | 96-well Strip Plate |
| Produkttype | Solid Phase Sandwich ELISA |
| Til brug med (applikation) | ELISA |
| Gen-id (Entrez) | 3002 |
| Gen symbol | GZMB |
| Sættets indhold | Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) |
| Mængde | 1 Kit (for 5 Plates) |
| Opbevaringskrav | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Vis mere |