Få mere at vide
Invitrogen™ Human GPN3 (aa 14-92) Control Fragment Recombinant Protein
Beskrivelse
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61591 (PA5-61591. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Tekniske data
Tekniske data
| Adgangsnummer | Q9UHW5 |
| Koncentration | ≥5.0 mg/mL |
| Til brug med (applikation) | Blocking Assay, Control |
| Formulering | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-id (Entrez) | 51184 |
| Navn | Human GPN3 (aa 14-92) Control Fragment |
| Mængde | 100 μL |
| Regulatorisk status | RUO |
| Gene Alias | AD-009; ATP binding domain 1 family, member C; ATPBD1C; ATP-binding domain 1 family member C; GPN3; GPN-loop GTPase 3; protein x 0004; UNQ1876/PRO4319 |
| Fælles navn | GPN3 |
| Vis mere |