Learn More
Abnova™ Human GMPPB Full-length ORF (AAH01141.1, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029925-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is thought to encode a GDP-mannose pyrophosphorylase. The encoded protein catalyzes the conversion of mannose-1-phosphate and GTP to GDP-mannose, a reaction involved in the production of N-linked oligosaccharides. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]
Sequence: MKALILVGGYGTRLRPLTLSTPKPLVDFCNKPILLHQVEALAAAGVDHVILAVSYMSQVLEKEMKAQEQRLGIRISMSHEEEPLGTAGPLALARDLLSETADPFFVLNSDVICDFPFQAMVQFHRHHGQEGSILVTKVEEPSKYGVVVCEADTGRIHRFVEKPQVFVSNKINAGMYILSPAVLRRIQLQPTSIEKEVFPIMAKEGQLYAMELQGFWMDIGQPKDFLTGMCLFLQSLRQKQPERLCSGPGIVGNVLVDPSARIGQNCSIGPNVSLGPGVVVEDGVCIRRCTVLRDARIRSHSWLESCIVGWRCRVGQWVRMENVTVLGEDVIVNDELYLNGASVLPHKSIGESVPEPRIIMSpecifications
AAH01141.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1851 | |
GMPPB | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
29925 | |
GMPPB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKALILVGGYGTRLRPLTLSTPKPLVDFCNKPILLHQVEALAAAGVDHVILAVSYMSQVLEKEMKAQEQRLGIRISMSHEEEPLGTAGPLALARDLLSETADPFFVLNSDVICDFPFQAMVQFHRHHGQEGSILVTKVEEPSKYGVVVCEADTGRIHRFVEKPQVFVSNKINAGMYILSPAVLRRIQLQPTSIEKEVFPIMAKEGQLYAMELQGFWMDIGQPKDFLTGMCLFLQSLRQKQPERLCSGPGIVGNVLVDPSARIGQNCSIGPNVSLGPGVVVEDGVCIRRCTVLRDARIRSHSWLESCIVGWRCRVGQWVRMENVTVLGEDVIVNDELYLNGASVLPHKSIGESVPEPRIIM | |
RUO | |
GMPPB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |