missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Human Glypican 4 (NP_001439) VersaClone cDNA
Beskrivelse
The Human Glypican 4 (NP_001439) VersaClone cDNA clone contains the complete ORF for Glypican 4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Glypican 4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Tekniske data
Tekniske data
| Gene | hGPC4 |
| Adgangsnummer | NP_001439 |
| Koncentration | 10 μg at 0.2 μg/μL |
| Formulering | cDNA is provided in 10 mM Tris-Cl |
| Indsæt type | 1684bp |
| pH | 8.5 |
| Gen-id (Entrez) | 2239 |
| Indhold og opbevaring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Arter | Human |
| Mængde | 10 μg |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?