missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GLYATL2 Full-length ORF (AAH16789.1, 1 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00219970-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MLVLHNSQKLQILYKSLEKSIPESIKVYGAIFNIKDKNPFNMEVLVDAWPDYQIVITRPQKQEMKDDQDHYTNTYHIFTKASDKLEEVLSYSNVISWEQTLQIQGCQEGLDEAIRKVATSKSVQVDYMKTILFIPELPKKHKTSSNDKMELFEVDDDNKKGNFSNMFIDASHAGLVNEHWAFGKNERSLKYIERCLQDFLGFGVLGPEGQLVSWIVMEQSCELRMGYTVPKYRHQGNMLQIGYHLEKYLSQKEIPFYFHVADNNEKSLQALNNLGFKICPCGWHQWKCTPKKYCSpecifications
AAH16789.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BXMAS2-10/GATF-B/MGC24009 | |
GLYATL2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
219970 | |
GLYATL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLVLHNSQKLQILYKSLEKSIPESIKVYGAIFNIKDKNPFNMEVLVDAWPDYQIVITRPQKQEMKDDQDHYTNTYHIFTKASDKLEEVLSYSNVISWEQTLQIQGCQEGLDEAIRKVATSKSVQVDYMKTILFIPELPKKHKTSSNDKMELFEVDDDNKKGNFSNMFIDASHAGLVNEHWAFGKNERSLKYIERCLQDFLGFGVLGPEGQLVSWIVMEQSCELRMGYTVPKYRHQGNMLQIGYHLEKYLSQKEIPFYFHVADNNEKSLQALNNLGFKICPCGWHQWKCTPKKYC | |
RUO | |
GLYATL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |