missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GIF Partial ORF (NP_005133.2, 318 a.a. - 417 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

2735.00 DKK - 4145.00 DKK

Specifications

Accession Number NP_005133.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2694
Molecular Weight (g/mol) 36.74kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16198664
View Documents
Abnova™
H00002694-Q01.25ug
25 ug This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16188664
View Documents
Abnova™
H00002694-Q01.10ug
10 ug This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This gene is a member of the cobalamin transport protein family. It encodes a glycoprotein secreted by parietal cells of the gastric mucosa and is required for adequate absorption of vitamin B12. Vitamin B12 is necessary for erythrocyte maturation and mutations in this gene may lead to congenital pernicious anemia. [provided by RefSeq]

Sequence: INNQLRGVELLFNETINVSVKSGSVLLVVLEEAQRKNPMFKFETTMTSWGLVVSSINNIAENVNHKTYWQFLSGVTPLNEGVADYIPFNHEHITANFTQY
Specifications

Specifications

NP_005133.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IF/IFMH/INF/TCN3
GIF
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
2694
GIF (Human) Recombinant Protein (Q01)
INNQLRGVELLFNETINVSVKSGSVLLVVLEEAQRKNPMFKFETTMTSWGLVVSSINNIAENVNHKTYWQFLSGVTPLNEGVADYIPFNHEHITANFTQY
RUO
GIF
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human GIF Partial ORF (NP_005133.2, 318 a.a. - 417 a.a.) Recombinant Protein with GST-tag at N-terminal

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.