missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Human GHSR (NP_940799) VersaClone cDNA

Product Code. 18461940
Change view
Click to view available options
Mængde:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18461940 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18461940 Supplier R&D Systems™ Supplier No. RDC0404
  • ONLINE EXCLUSIVE PRICE
    2605.00 DKK
    2460.15 DKK / 10µg
    Spare 144.85 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Growth Hormone Secretagogue Receptor

The Human GHSR (NP_940799) VersaClone cDNA clone contains the complete ORF for Ghrelin Receptor/GHSR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Ghrelin Receptor/GHSR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Gene hGHSR
Adgangsnummer NP_940799
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10mM Tris-Cl, pH 8.5
Indsæt type 1114bp
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Human
Produktlinje R&D Systems
Mængde 10 μg

For Reasearch Use Only; Not for Use in Humans

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.