Learn More
Abnova™ Human GGT1 Full-length ORF (NP_005256.2, 1 a.a. - 569 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_005256.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2678 |
Molecular Weight (g/mol) | 87.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138177
|
Abnova™
H00002678-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16148177
|
Abnova™
H00002678-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The enzyme encoded by this gene catalyzes the transfer of the glutamyl moiety of glutathione to a variety of amino acids and dipeptide acceptors. The enzyme is composed of a heavy chain and a light chain, which are derived from a single precursor protein, and is present in tissues involved in absorption and secretion. This enzyme is a member of the gamma-glutamyltransferase protein family, of which many members have not yet been fully characterized and some of which may represent pseudogenes. This gene is classified as type I gamma-glutamyltransferase. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Sequence: MKKKLVVLGLLAVVLVLVIVGLCLWLPSASKEPDNHVYTRAAVAADAKQCSKIGRDALRDGGSAVDAAIAALLCVGLMNAHSMGIGGGLFLTIYNSTTRKAEVINAREVAPRLAFATMFNSSEQSQKGGLSVAVPGEIRGYELAHQRHGRLPWARLFQPSIQLARQGFPVGKGLAAALENKRTVIEQQPVLCEVFCRDRKVLREGERLTLPQLADTYETLAIEGAQAFYNGSLTAQIVKDIQAAGGIVTAEDLNNYRAELIEHPLNISLGDVVLYMPSAPLSGPVLALILNILKGYNFSRESVESPEQKGLTYHRIVEAFRFAYAKRTLLGDPKFVDVTEVVRNMTSEFFAAQLRAQISDDTTHPISYYKPEFYTPDDGGTAHLSVVAEDGSAVSATSTINLYFGSKVRSPVSGILFNNEMDDFSSPSITNEFGVPPSPANFIQPGKQPLSSMCPTIMVGQDGQVRMVVGAAGGTQITTATALAIIYNLWFGYDVKRAVEEPRLHNQLLPNVTTVERNIDQAVTAALETRHHHTQIASTFIAVVQAIVRTAGGWAAASDSRKGGEPAGYSpecifications
NP_005256.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
87.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKKKLVVLGLLAVVLVLVIVGLCLWLPSASKEPDNHVYTRAAVAADAKQCSKIGRDALRDGGSAVDAAIAALLCVGLMNAHSMGIGGGLFLTIYNSTTRKAEVINAREVAPRLAFATMFNSSEQSQKGGLSVAVPGEIRGYELAHQRHGRLPWARLFQPSIQLARQGFPVGKGLAAALENKRTVIEQQPVLCEVFCRDRKVLREGERLTLPQLADTYETLAIEGAQAFYNGSLTAQIVKDIQAAGGIVTAEDLNNYRAELIEHPLNISLGDVVLYMPSAPLSGPVLALILNILKGYNFSRESVESPEQKGLTYHRIVEAFRFAYAKRTLLGDPKFVDVTEVVRNMTSEFFAAQLRAQISDDTTHPISYYKPEFYTPDDGGTAHLSVVAEDGSAVSATSTINLYFGSKVRSPVSGILFNNEMDDFSSPSITNEFGVPPSPANFIQPGKQPLSSMCPTIMVGQDGQVRMVVGAAGGTQITTATALAIIYNLWFGYDVKRAVEEPRLHNQLLPNVTTVERNIDQAVTAALETRHHHTQIASTFIAVVQAIVRTAGGWAAASDSRKGGEPAGY | |
RUO | |
GGT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
2678 | |
GGT1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD224/D22S672/D22S732/GGT/GTG/MGC96892/MGC96904/MGC96963 | |
GGT1 | |
Recombinant | |
wheat germ expression system |