missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GFPT1 Full-length ORF (AAH45641.1, 1 a.a. - 681 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002673-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135431

  • 4040.00 DKK / 25µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes the first and rate-limiting enzyme of the hexosamine pathway and controls the flux of glucose into the hexosamine pathway. The product of this gene catalyzes the formation of glucosamine 6-phosphate. [provided by RefSeq]

Sequence: MCGIFAYLNYHVPRTRREILETLIKGLQRLEYRGYDSAGVGFDGGNDKDWEANACKIQLIKKKGKVKALDEEVHKQQDMDLDIEFDVHLGIAHTRWATHGEPSPVNSHPQRSDKNNEFIVIHNGIITNYKDLKKFLESKGYDFESETDTETIAKLVKYMYDNRESQDTSFTTLVERVIQQLEGAFALVFKSVHFPGQAVGTRRGSPLLIGVRSEHKLSTDHIPILYRTGKDKKGSCNLSRVDSTTCLFPVEEKAVEYYFASDASAVIEHTNRVIFLEDDDVAAVVDGRLSIHRIKRTAGDHPGRAVQTLQMELQQIMKGNFSSFMQKEIFEQPESVVNTMRGRVNFDDYTVNLGGLKDHIKEIQRCRRLILIACGTSYHAGVATRQVLEELTELPVMVELASDFLDRNTPVFRDDVCFFLSQSGETADTLMGLRYCKERGALTVGITNTVGSSISRETDCGVHINAGPEIGVASTKAYTSQFVSLVMFALMMCDDRISMQERRKEIMLGLKRLPDLIKEVLSMDDEIQKLATELYHQKSVLIMGRGYHYATCLEGALKIKEITYMHSEGILAGELKHGPLALVDKLMPVIMIIMRDHTYAKCQNALQQVVARQGRPVVICDKEDTETIKNTKRTIKVPHSVDCLQGILSVIPLQLLAFHLAVLRGYDVDFPRNLAKSVTVE
Specifications

Specifications

AAH45641.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
103.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GFA/GFAT/GFAT1/GFAT1m/GFPT
GFPT1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
2673
GFPT1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MCGIFAYLNYHVPRTRREILETLIKGLQRLEYRGYDSAGVGFDGGNDKDWEANACKIQLIKKKGKVKALDEEVHKQQDMDLDIEFDVHLGIAHTRWATHGEPSPVNSHPQRSDKNNEFIVIHNGIITNYKDLKKFLESKGYDFESETDTETIAKLVKYMYDNRESQDTSFTTLVERVIQQLEGAFALVFKSVHFPGQAVGTRRGSPLLIGVRSEHKLSTDHIPILYRTGKDKKGSCNLSRVDSTTCLFPVEEKAVEYYFASDASAVIEHTNRVIFLEDDDVAAVVDGRLSIHRIKRTAGDHPGRAVQTLQMELQQIMKGNFSSFMQKEIFEQPESVVNTMRGRVNFDDYTVNLGGLKDHIKEIQRCRRLILIACGTSYHAGVATRQVLEELTELPVMVELASDFLDRNTPVFRDDVCFFLSQSGETADTLMGLRYCKERGALTVGITNTVGSSISRETDCGVHINAGPEIGVASTKAYTSQFVSLVMFALMMCDDRISMQERRKEIMLGLKRLPDLIKEVLSMDDEIQKLATELYHQKSVLIMGRGYHYATCLEGALKIKEITYMHSEGILAGELKHGPLALVDKLMPVIMIIMRDHTYAKCQNALQQVVARQGRPVVICDKEDTETIKNTKRTIKVPHSVDCLQGILSVIPLQLLAFHLAVLRGYDVDFPRNLAKSVTVE
RUO
GFPT1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers