missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gibco™ Human GDF-15/MIC-1 Recombinant Protein, PeproTech®
Klik for at se tilgængelige muligheder
Mængde:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Beskrivelse
Recombinant Human GDF-15/MIC-1 is a disulfide linked homodimeric protein consisting of two 112 amino acid polypeptide chains. The calculated molecular weight of Recombinant Human GDF-15/MIC-1 is 24.6 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
GDF-15 belongs to the TGF-beta cytokine family, whose members play an important role during prenatal development and postnatal growth, and the remodeling and maintenance of a variety of tissues and organs. GDF-15 is expressed predominantly in the placenta and, to a much lesser extent, in various other tissues. The presence of GDF-15 in amniotic fluid and its elevated levels in the sera of pregnant women suggest GDF-15's involvement in gestation and embryonic development. GDF-15 generally exerts tumor suppressive activities and is one of the predominant factors produced and secreted in response to activation of the p53 pathway. Interestingly, the serum level of GDF-15 is positively correlated with neoplastic progression of several tumor types, including certain colorectal, pancreatic, and prostate cancers.
Tekniske data
Tekniske data
| Adgangsnummer | Q99988 |
| Til brug med (applikation) | Functional Assay |
| Formulering | protein with no preservative |
| Gen-id (Entrez) | 9518 |
| Molekylvægt (g/mol) | 24.6 kDa |
| Navn | Human GDF-15/MIC-1 |
| Mængde | 1 mg |
| Kilde | CHO cells |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <1 EU/ μg |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion