missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Human GASP-1 Recombinant Protein, PeproTech®

Artikelnummer. 17820493
Klik for at se tilgængelige muligheder
Mængde:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 17820493

Brand: Gibco™ 120415UG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Recombinant Human GASP-1 is a 542 amino acid protein that migrates at an apparent molecular weight of approximately 55-66 kDa by SDS-PAGE analysis under non-reducing conditions. The calculated molecular weight of Recombinant Human GASP-1 is 59.9 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Growth and differentiation factor-associated serum protein-1 (GASP-1) is a secreted inhibitory TGF-beta binding protein that contains multiple protease inhibitor structural domains. It is expressed primarily in the ovary, testis, and brain, and can act as a potent soluble inhibitor of myostatin and GDF-11, but not Activin A. The GASP-1 gene encodes a 571 amino acid protein that contains a 29 amino acid secretion signal sequence, and multiple identifiable structural features, including a WAP domain, a follistatin/Kazal domain, an immunoglobulin domain, two tandem Kunitz domains, and a netrin domain.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer Q8TEU8
Til brug med (applikation) Functional Assay
Formulering protein with no preservative
Gen-id (Entrez) 124857
Molekylvægt (g/mol) 59.9 kDa
Navn Human GASP-1
Mængde 5 μg
Kilde CHO cells
Regulatorisk status RUO
Endotoksin koncentration <1 EU/ μg
Gene Alias 2610304F08Rik; AY100450; Gasp1; GASP-1; Growth and differentiation factor-associated serum protein 1; hGASP-1; mGASP-1; multivalent protease inhibitor protein; RGD1305361; UNQ9235/PRO31996; WAP four-disulfide core domain 20 B; WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing-related protein; WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2; WAP, FS, Ig, two KU and NTR module related protein; WAP, Kazal, immunoglobulin, Kunitz and NTR domain-containing protein 2; WAP, kazal, immunoglobulin, kunitz and NTR domain-containing protein 2; WAP, Kazal, immunoglobulin, Kunitz and NTR domain-containing protein 2; WFDC20B; WFIKKN2; WFIKKN-related protein; WFIKKNRP
Fælles navn GASP1
Gen symbol WFIKKN2
Biologisk aktivitet Determined by its ability to inhibit human Myostatin (GDF-8) activity in MCP-11 cells. The ED50 for this effect is 0.0025-0.0040 ug/ml in the presence of 5 ng/ml of human Myostatin (GDF-8).
Konjugeret Unconjugated
Rekombinant Recombinant
Sekvens LPPIRYSHAG ICPNDMNPNL WVDAQSTCRR ECETDQECET YEKCCPNVCG TKSCVAARYM DVKGKKGPVG MPKEATCDHF MCLQQGSECD IWDGQPVCKC KDRCEKEPSF TCASDGLTYY NRCYMDAEAC SKGITLAVVT CRYHFTWPNT SPPPPETTMH PTTASPETPE LDMAAPALLN NPVHQSVTMG ETVSFLCDVV GRPRPEITWE KQLEDRENVV MRPNHVRGNV VVTNIAQLVI YNAQLQDAGI YTCTARNVAG VLRADFPLSV VRGHQAAATS ESSPNGTAFP AAECLKPPDS EDCGEEQTRW HFDAQANNCL TFTFGHCHRN LNHFETYEAC MLACMSGPLA ACSLPALQGP CKAYAPRWAY NSQTGQCQSF VYGGCEGNGN NFESREACEE SCPFPRGNQR CRACKPRQKL VTSFCRSDFV ILGRVSELTE EPDSGRALVT VDEVLKDEKM GLKFLGQEPL EVTLLHVDWA CPCPNVTVSE MPLIIMGEVD GGMAMLRPDS FVGASSARRV RKLREVMHKK TCDVLKEFLG LH
Indhold og opbevaring -20°C
Udtrykssystem CHO cells
Form Lyophilized
Renhed eller kvalitetsklasse ≥ 95% by SDS-PAGE gel and HPLC analyses.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.