missing translation for 'onlineSavingsMsg'
Få mere at vide

Human Gas6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. 18191691
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18191691 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18191691 Leverandør R&D Systems Leverandørnr. BAF885

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Goat Polyclonal Antibody has been used in 6 publications

Gas6 Polyclonal specifically detects Gas6 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifica

Antigen Gas6
Applikationer Western Blot
Klassifikation Polyclonal
Koncentration LYOPH
Konjugeret Biotin
Fortynding Western Blot 0.1 ug/mL
Genadgangsnr. NP_000811
Gene Alias AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6
Gensymboler GAS6
Værtsarter Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Gas6 Asp118-Ala678 Accession # NP_000811
Oprensningsmetode Affinity Purified
Mængde 50 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 2621
Testspecificitet Detects human Gas6 in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse Gas6 is observed.
Rekonstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Målarter Human
Indhold og opbevaring Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.