Learn More
Abnova™ Human GALNT7 Full-length ORF (AAH46129.1, 1 a.a. - 657 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAH46129.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51809 |
Molecular Weight (g/mol) | 101.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16150373
|
Abnova™
H00051809-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140373
|
Abnova™
H00051809-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes GalNAc transferase 7, a member of the GalNAc-transferase family. The enzyme encoded by this gene controls the initiation step of mucin-type O-linked protein glycosylation and transfer of N-acetylgalactosamine to serine and threonine amino acid residues. This enzyme is a type II transmembrane protein and shares common sequence motifs with other family members. Unlike other family members, this enzyme shows exclusive specificity for partially GalNAc-glycosylated acceptor substrates and shows no activity with non-glycosylated peptides. This protein may function as a follow-up enzyme in the initiation step of O-glycosylation. [provided by RefSeq]
Sequence: MRLKIGFILRSLLVVGSFLGLVVLWSSLTPRPDDPSPLSRMREDRDVNDPMPNRGGNGLAPGEDRFKPVVPWPHVEGVEVDLESIRRINKAKNEQEHHAGGDSQKDIMQRQYLTFKPQTFTYHDPVLRPGILGNFEPKEPEPPGVVGGPGEKAKPLVLGPEFKQAIQASIKEFGFNMVASDMISLDRSVNDLRQEECKYWHYDENLLTSSVVIVFHNEGWSTLMRTVHSVIKRTPRKYLAEIVLIDDFSNKEHLKEKLDEYIKLWNGLVKVFRNERREGLIQARSIGAQKAKLGQVLIYLDAHCEVAVNWYAPLVAPISKDRTICTVPLIDVINGNTYEIIPQGGGDEDGYARGAWDWSMLWKRVPLTPQEKRLRKTKTEPYRSPAMAGGLFAIEREFFFELGLYDPGLQIWGGENFEISYKIWQCGGKLLFVPCSRVGHIYRLEGWQGNPPPIYVGSSPTLKNYVRVVEVWWDEYKDYFYASRPESQALPYGDISELKKFREDHNCKSFKWFMEEIAYDITSHYPLPPKNVDWGEIRGFETAYCIDSMGKTNGGFVELGPCHRMGGNQLFRINEANQLMQYDQCLTKGADGSKVMITHCNLNEFKEWQYFKNLHRFTHIPSGKCLDRSEVLHQVFISNCDSSKTTQKWEMNNIHSVSpecifications
AAH46129.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
101.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MRLKIGFILRSLLVVGSFLGLVVLWSSLTPRPDDPSPLSRMREDRDVNDPMPNRGGNGLAPGEDRFKPVVPWPHVEGVEVDLESIRRINKAKNEQEHHAGGDSQKDIMQRQYLTFKPQTFTYHDPVLRPGILGNFEPKEPEPPGVVGGPGEKAKPLVLGPEFKQAIQASIKEFGFNMVASDMISLDRSVNDLRQEECKYWHYDENLLTSSVVIVFHNEGWSTLMRTVHSVIKRTPRKYLAEIVLIDDFSNKEHLKEKLDEYIKLWNGLVKVFRNERREGLIQARSIGAQKAKLGQVLIYLDAHCEVAVNWYAPLVAPISKDRTICTVPLIDVINGNTYEIIPQGGGDEDGYARGAWDWSMLWKRVPLTPQEKRLRKTKTEPYRSPAMAGGLFAIEREFFFELGLYDPGLQIWGGENFEISYKIWQCGGKLLFVPCSRVGHIYRLEGWQGNPPPIYVGSSPTLKNYVRVVEVWWDEYKDYFYASRPESQALPYGDISELKKFREDHNCKSFKWFMEEIAYDITSHYPLPPKNVDWGEIRGFETAYCIDSMGKTNGGFVELGPCHRMGGNQLFRINEANQLMQYDQCLTKGADGSKVMITHCNLNEFKEWQYFKNLHRFTHIPSGKCLDRSEVLHQVFISNCDSSKTTQKWEMNNIHSV | |
RUO | |
GALNT7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51809 | |
GALNT7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GALNAC-T7/GalNAcT7 | |
GALNT7 | |
Recombinant | |
wheat germ expression system |