missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gibco™ Human Galectin-3 Recombinant Protein, PeproTech®
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Beskrivelse
Recombinant Human Galectin-3 is a globular 26.0 kDa protein containing 249 amino acid residues, but no disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Galactose-specific lectin which binds IgE. May mediate with the alpha-3, beta-1 integrin the stimulation by CSPG4 of endothelial cells migration. Together with DMBT1, required for terminal differentiation of columnar epithelial cells during early embryogenesis. In the nucleus: acts as a pre-mRNA splicing factor. Involved in acute inflammatory responses including neutrophil activation and adhesion, chemoattraction of monocytes macrophages, opsonization of apoptotic neutrophils, and activation of mast cells.
Tekniske data
Tekniske data
| Adgangsnummer | P17931 |
| Til brug med (applikation) | ELISA, Functional Assay, Western Blot |
| Formulering | protein with no preservative |
| Gen-id (Entrez) | 3958 |
| Molekylvægt (g/mol) | 26 kDa |
| Navn | Human Galectin-3 |
| Mængde | 500 μg |
| Kilde | E. coli |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <1 EU/ μg |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion