missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Galectin-1 DuoSet ELISA
Description
Includes
Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
The Human Galectin-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Galectin-1.
Specifications
Specifications
| Analyseområde | 0.312 to 20ng/mL |
| Detekterbare analytter | Galectin-1 |
| Assay | Solid Phase ELISA |
| Konjugeret | Biotin |
| Krydsreaktivitet | <50%cross-species reactivity observed with species tested |
| Immunoassay-kitformat | 96-well Strip Plate |
| Produkttype | ELISA Kit |
| Prøvetype | Cell Culture Supernatant |
| Tilstrækkelig til | To run ELISAs on at least 5 x 96-well plates |
| Målarter | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?