Learn More
Abnova™ Human GABPB2 Full-length ORF (NP_005245.2, 1 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002553-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes the GA-binding protein transcription factor, beta subunit. This protein forms a tetrameric complex with the alpha subunit, and stimulates transcription of target genes. The encoded protein may be involved in activation of cytochrome oxidase expression and nuclear control of mitochondrial function. The crystal structure of a similar protein in mouse has been resolved as a ternary protein complex. Multiple transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MSLVDLGKKLLEAARAGQDDEVRILMANGAPFTTDWLGTSPLHLAAQYGHYSTTEVLLRAGVSRDARTKVDRTPLHMAASEGHASIVEVLLKHGADVNAKDMLKMTALHWATEHNHQEVVELLIKYGADVHTQSKFCKTAFDISIDNGNEDLAEILQIAMQNQINTNPESPDTVTIHAATPQFIIGPGGVVNLTGLVSSENSSKATDETGVSAVQFGNSSTSVLATLAALAEASAPLSNSSETPVVATEEVVTAESVDGAIQQVVSSGGQQVITIVTDGIQLGNLHSIPTSGIGQPIIVTMPDGQQVLTVPATDIAEETVISEEPPAKRQCIEIIENRVESAEIEEREALQKQLDEANREAQKYRQQLLKKEQEAEAYRQKLEAMTRLQTNKEAVSpecifications
NP_005245.2 | |
Liquid | |
2553 | |
GABPB2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSLVDLGKKLLEAARAGQDDEVRILMANGAPFTTDWLGTSPLHLAAQYGHYSTTEVLLRAGVSRDARTKVDRTPLHMAASEGHASIVEVLLKHGADVNAKDMLKMTALHWATEHNHQEVVELLIKYGADVHTQSKFCKTAFDISIDNGNEDLAEILQIAMQNQINTNPESPDTVTIHAATPQFIIGPGGVVNLTGLVSSENSSKATDETGVSAVQFGNSSTSVLATLAALAEASAPLSNSSETPVVATEEVVTAESVDGAIQQVVSSGGQQVITIVTDGIQLGNLHSIPTSGIGQPIIVTMPDGQQVLTVPATDIAEETVISEEPPAKRQCIEIIENRVESAEIEEREALQKQLDEANREAQKYRQQLLKKEQEAEAYRQKLEAMTRLQTNKEAV | |
RUO | |
GABPB1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BABPB2/E4TF1/E4TF1-47/E4TF1-53/E4TF1B/GABPB/GABPB2/NRF2B1/NRF2B2 | |
GABPB1 | |
Yes | |
wheat germ expression system |