missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Furin DuoSet ELISA
Description
The Human Furin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Furin.
Specifications
Specifications
| Mængde | 1 sæt (til 15 tallerkener) |
| Arter | Human |
| Konjugeret | Biotin |
| Analyseområde | 0.31 to 20 ng/mL |
| Til brug med (applikation) | ELISA |
| Gen-id (Entrez) | 5045 |
| Synonym | Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FUR, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACE, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PC, PCSK3, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1 |
| Indhold og opbevaring | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?