missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXG1 Partial ORF (CAA52241.1, 164 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002290-Q03.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16053995

  • 4315.00 DKK / 25µg
Estimated Shipment: 27-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon. [provided by RefSeq]

Sequence: SPFLSLHHPRASSTLSYNGTTSAYPSHPMPYSSVLTQNSLGNNHSFSTANGLSVDRLVNGE
Specifications

Specifications

CAA52241.1
Liquid
2290
FOXG1 (Human) Recombinant Protein (Q03)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BF1/BF2/FHKL3/FKH2/FKHL1/FKHL2/FKHL3/FKHL4/FOXG1A/FOXG1B/FOXG1C/HBF-1/HBF-2/HBF-3/HBF-G2/HBF2/HFK1/HFK2/HFK3/KHL2/QIN
FOXG1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.45kDa
12.5% SDS-PAGE Stained with Coomassie Blue
SPFLSLHHPRASSTLSYNGTTSAYPSHPMPYSSVLTQNSLGNNHSFSTANGLSVDRLVNGE
RUO
FOXG1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers