missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXD4L4 Partial ORF (NP_954714.2, 317 a.a. - 416 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00349334-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132877

  • 2735.00 DKK / 10µg
Estimated Shipment: 24-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: HREADASLSALRVLCKGSGERVQGLRRVCPRPRGATATCSSDHQACCIPKPLPLCCKCPPPLLLGQFCSNSSSIRRTAPTAALPPRARCWAGTCRPRRRC
Specifications

Specifications

NP_954714.2
Liquid
349334
FOXD4L4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FOXD4b/bA460E7.2
FOXD4L4
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HREADASLSALRVLCKGSGERVQGLRRVCPRPRGATATCSSDHQACCIPKPLPLCCKCPPPLLLGQFCSNSSSIRRTAPTAALPPRARCWAGTCRPRRRC
RUO
FOXD4L4
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers