missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FOXD4L4 Partial ORF (NP_954714.2, 317 a.a. - 416 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00349334-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: HREADASLSALRVLCKGSGERVQGLRRVCPRPRGATATCSSDHQACCIPKPLPLCCKCPPPLLLGQFCSNSSSIRRTAPTAALPPRARCWAGTCRPRRRCSpecifications
NP_954714.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HREADASLSALRVLCKGSGERVQGLRRVCPRPRGATATCSSDHQACCIPKPLPLCCKCPPPLLLGQFCSNSSSIRRTAPTAALPPRARCWAGTCRPRRRC | |
RUO | |
FOXD4L4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
349334 | |
FOXD4L4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FOXD4b/bA460E7.2 | |
FOXD4L4 | |
Recombinant | |
wheat germ expression system |