Learn More
Abnova™ Human FLOT1 Partial ORF (NP_005794.1, 126 a.a. - 225 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010211-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined. [provided by RefSeq]
Sequence: KFSEQVFKVASSDLVNMGISVVSYTLKDIHDDQDYLHSLGKARTAQVQKDARIGEAEAKRDAGIREAKAKQEKVSAQYLSEIEMAKAQRDYELKKAAYDISpecifications
NP_005794.1 | |
Liquid | |
10211 | |
FLOT1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLOT1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KFSEQVFKVASSDLVNMGISVVSYTLKDIHDDQDYLHSLGKARTAQVQKDARIGEAEAKRDAGIREAKAKQEKVSAQYLSEIEMAKAQRDYELKKAAYDI | |
RUO | |
FLOT1 | |
Yes | |
wheat germ expression system |