missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FGFR1OP2 Partial ORF (NP_056448, 62 a.a. - 169 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026127-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135326

  • 2735.00 DKK / 10µg
Estimated Shipment: 12-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: RSTLVMGIQQENRQIRELQQENKELRTSLEEHQSALELIMSKYREQMFRLLMASKKDDPGIIMKLKEQHSKIDMVHRNKSEGFFLDASRHILEAPQHGLERRHLEANQ
Specifications

Specifications

NP_056448
Liquid
26127
FGFR1OP2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp564O1863/HSPC123-like
FGFR1OP2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RSTLVMGIQQENRQIRELQQENKELRTSLEEHQSALELIMSKYREQMFRLLMASKKDDPGIIMKLKEQHSKIDMVHRNKSEGFFLDASRHILEAPQHGLERRHLEANQ
RUO
FGFR1OP2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers