missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-9, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
1610.00 DKK - 31830.00 DKK
Specifications
Accession Number | P31371 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2254 |
Molecular Weight (g/mol) | 23.2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17843573
|
Gibco™
AF-100-23-100UG |
100 μg |
6570.00 DKK
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17853013
|
Gibco™
AF-100-23-1MG |
1 mg |
31830.00 DKK
1mg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17893233
|
Gibco™
AF-100-23-20UG |
20 μg |
1610.00 DKK
20µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17803773
|
Gibco™
AF-100-23-500UG |
500 μg |
19070.00 DKK
500µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis.Specifications
P31371 | |
protein with no preservative | |
23.2 | |
E. coli | |
RUO | |
Eks; elbow knee synostosis; FGF; Fgf9; FGF-9; Fibroblast growth factor; Fibroblast growth factor 9; fibroblast growth factor 9 (glia-activating factor); GAF; glia activating factor; glia-activating factor; HBFG-9; HBGF-9; Heparin-binding growth factor 9; M-FGF-9; SYNS3 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
2254 | |
Human FGF-9, Animal-Free | |
-20°C | |
<0.1 EU/μg | |
FGF9 | |
Unconjugated | |
PLGEVGNYFG VQDAVPFGNV PVLPVDSPVL LSDHLGQSEA GGLPRGPAVT DLDHLKGILR RRQLYCRTGF HLEIFPNGTI QGTRKDHSRF GILEFISIAV GLVSIRGVDS GLYLGMNEKG ELYGSEKLTQ ECVFREQFEE NWYNTYSSNL YKHVDTGRRY YVALNKDGTP REGTRTKRHQ KFTHFLPRPV DPDKVPELYK DILSQS | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |