Få mere at vide
Gibco™ Human FGF-16 Recombinant Protein
Beskrivelse
Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in 0.5X phosphate buffered saline to a concentration of 0.1-1.0 mg/mL. Further dilutions should be made in aqueous buffers supplemented with carrier protein, such as 0.1-1.0% BSA. Storage: This lyophilized preparation is stable at 2°C to 8°C, but should be kept at -20°C for long term storage, preferably desiccated. Upon reconstitution, the preparation is stable for up to one week at 2°C to 8°C. For maximal stability, apportion the reconstituted preparation into working aliquots and store at -20°C. Avoid repeated freeze/thaw cycles.
Tekniske data
Tekniske data
| Adgangsnummer | O43320 |
| Til brug med (applikation) | Bioactivity |
| Formulering | 5mM sodium phosphate with 25mM NaCl and no preservative; pH 7.2 |
| Gen-id (Entrez) | 8823 |
| Molekylvægt (g/mol) | 23.7 kDa |
| Navn | Human FGF-16 |
| pH-område | 7.2 |
| Oprensningsmetode | Purified |
| Mængde | 25 μg |
| Opbevaringskrav | -20°C |
| Vis mere |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.