Learn More
Abnova™ Human FBXL3 Partial ORF (NP_036290, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_036290 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26224 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138182
|
Abnova™
H00026224-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16128182
|
Abnova™
H00026224-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats and is localized in the nucleus. [provided by RefSeq]
Sequence: MKRGGRDSDRNSSEEGTAEKSKKLRTTNEHSQTCDWGNLLQDIILQVFKYLPLLDRAHASQVCRNWNQVFHMPDLWRCFEFELNQPATSYLKATHPELIKSpecifications
NP_036290 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBL3/FBL3A/FBXL3A | |
FBXL3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
26224 | |
FBXL3 (Human) Recombinant Protein (Q01) | |
MKRGGRDSDRNSSEEGTAEKSKKLRTTNEHSQTCDWGNLLQDIILQVFKYLPLLDRAHASQVCRNWNQVFHMPDLWRCFEFELNQPATSYLKATHPELIK | |
RUO | |
FBXL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |