missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human F-Spondin/SPON1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3135-SP
This item is not returnable.
View return policy
Description
F-Spondin/SPON1 Polyclonal specifically detects F-Spondin/SPON1 in Human samples. It is validated for Western Blot, Simple Western, Neutralization.
Specifications
F-Spondin/SPON1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
f-spondin, FSpondin, KIAA0762VSGP/F-spondin, MGC10724, SPON1, spondin 1, (f-spondin) extracellular matrix protein, spondin 1, extracellular matrix protein, spondin-1, Vascular smooth muscle cell growth-promoting factor, VSGP | |
Goat | |
Affinity Purified | |
RUO | |
10418 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Neutralization 3 ug/mL | |
Q9HCB6 | |
SPON1 | |
Mouse myeloma cell line NS0-derived recombinant human F-Spondin/SPON1 Phe29-Cys807 Accession # Q9HCB6 | |
25 μg | |
Primary | |
Detects human F-Spondin/SPON1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human F-Spondin/SPON1 Antibody, R&D Systems™ > 25μg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction